Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028711.3 | NP_082987.1 | 2 | 258 | ||
GenBank mRNA | AB106930.1 | - | 2 | 402 | ||
AK014394.1 | - | 2 | 259 | |||
AK021314.1 | - | 3 | 344 | |||
AK043831.1 | - | 2 | 458 | |||
AK054292.1 | - | 1 | 821 | |||
BC138993.1 | - | 2 | 265 | 8862015 | ||
BC138994.1 | - | 2 | 265 | 8862016 |