Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079814.1 | NP_001073282.1 | 4 | 276 | ||
NM_028720.2 | NP_082996.2 | 4 | 276 | |||
GenBank mRNA | AK014456.1 | - | 4 | 216 | ||
AK040841.1 | - | 4 | 226 | |||
AK140812.1 | - | 3 | 1243 | |||
AK145277.1 | - | 4 | 226 | |||
AK150349.1 | - | 4 | 220 | |||
AK152887.1 | - | 4 | 220 | |||
AK159476.1 | - | 4 | 230 | |||
AK181131.1 | - | 2 | 101 | |||
AK181230.1 | - | 2 | 101 | |||
AK183340.1 | - | 2 | 101 | |||
AK183942.1 | - | 2 | 101 | |||
AK186402.1 | - | 2 | 116 | |||
AK204179.1 | - | 2 | 101 | |||
AK214807.1 | - | 2 | 121 | |||
AK215028.1 | - | 2 | 101 | |||
AK215337.1 | - | 2 | 89 | |||
BC006893.1 | - | 4 | 210 | 3599477 | ||
BY135011.1 | - | 4 | 278 |