Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014568.1 | - | 4 | 898 | ||
AK020079.1 | - | 4 | 480 | |||
AK042581.1 | - | 4 | 884 | |||
AK046761.1 | - | 4 | 889 | |||
AK047599.1 | - | 4 | 913 | |||
AK050948.1 | - | 4 | 896 | |||
AK079935.1 | - | 4 | 897 | |||
AK157398.1 | - | 4 | 896 | |||
AK157956.1 | - | 5 | 801 | |||
AK164493.1 | - | 4 | 907 | |||
AK172043.1 | - | 4 | 902 | |||
BC024877.1 | - | 4 | 900 | |||
BC051109.1 | - | 4 | 869 | |||
BC096599.1 | - | 4 | 888 | |||
BC125432.1 | - | 4 | 895 | 40129947 | ||
BC132099.1 | - | 4 | 895 | 40130376 |