Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277292.1 | NP_001264221.1 | 3 | 553 | ||
NM_001277293.1 | NP_001264222.1 | 3 | 553 | |||
NM_001277294.1 | NP_001264223.1 | 3 | 410 | |||
NM_028736.2 | NP_083012.1 | 3 | 410 | |||
NM_130891.2 | NP_570961.1 | 2 | 329 | |||
NM_133442.2 | NP_597699.1 | 2 | 472 | |||
GenBank mRNA | AB051560.1 | - | 2 | 206 | ||
AB051561.1 | - | 3 | 287 | |||
AB051562.1 | - | 2 | 192 | |||
AK029845.1 | - | 3 | 208 | |||
AK039673.1 | - | 2 | 214 | |||
AK220553.1 | - | 3 | 267 | |||
BC067398.1 | - | 3 | 410 | 30096759 | ||
BC072632.1 | - | 3 | 553 | 6843802 |