Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifltd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028742.2 | NP_083018.1 | 4 | 452 | ||
GenBank mRNA | AK016641.1 | - | 4 | 452 | ||
AK052206.1 | - | 2 | 531 | |||
AY753326.1 | - | 2 | 287 | |||
BC053420.1 | - | 3 | 414 | 6705208 |