Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hvcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042489.2 | NP_001035954.1 | 4 | 629 | ||
NM_028752.3 | NP_083028.1 | 3 | 491 | |||
GenBank mRNA | AK002854.1 | - | 3 | 492 | ||
AK154880.1 | - | 4 | 521 | |||
AK155123.1 | - | 4 | 466 | |||
AK170422.1 | - | 4 | 490 | |||
BC021548.1 | - | 4 | 460 | 4913027 | ||
BY703352.1 | - | 3 | 485 | |||
CF734112.1 | - | 4 | 464 | |||
CF749152.1 | - | 4 | 629 |