Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028604.2 | NP_082880.2 | 9 | 990 | ||
GenBank mRNA | AF532979.1 | - | 9 | 990 | ||
AK010722.1 | - | 9 | 932 | |||
BC048703.1 | - | 7 | 665 | 6509593 |