Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028608.3 | NP_082884.1 | 5, 6 | 847 | ||
GenBank mRNA | AK010768.1 | - | 5, 6 | 779 | ||
AK137557.1 | - | 5, 6 | 794 | |||
AK170051.1 | - | 5, 6 | 794 | |||
BC025083.1 | - | 5, 6 | 771 | 3711757 | ||
BC098231.1 | - | 5, 6 | 821 | 3325823 | ||
BY540964.1 | - | 3, 4 | 145 |