Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028633.3 | NP_082909.1 | 10 | 1345 | ||
NM_207104.1 | NP_996987.1 | 7 | 997 | |||
GenBank mRNA | AK003871.1 | - | 7 | 968 | ||
AK050729.1 | - | 7 | 931 | |||
AY029337.1 | - | 10 | 1345 | |||
BC025464.1 | - | 10 | 1336 | 5351961 |