Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011053.2 | NP_035183.2 | 33 | 5176 | ||
GenBank mRNA | AK003899.1 | - | 2 | 358 | ||
AK129080.1 | - | 33 | 5168 | |||
BC010844.1 | - | 33 | 5149 | |||
BC038503.1 | - | 4 | 655 | |||
BC051231.1 | - | 1 | 503 | |||
BC055276.1 | - | 2 | 363 | |||
BC070468.1 | - | 33 | 5163 | 30628812 | ||
BC094244.1 | - | 13 | 2568 |