Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110129.1 | NP_001103599.1 | 5 | 267 | ||
NM_001110130.1 | NP_001103600.1 | 5 | 267 | |||
NM_028677.4 | NP_082953.1 | 5 | 267 | |||
GenBank mRNA | AK003179.1 | - | 5 | 245 | ||
AK005202.1 | - | 5 | 228 | |||
AK008394.1 | - | 5 | 245 | |||
AK014665.1 | - | 5 | 228 | |||
AK134995.1 | - | 5 | 247 | |||
AK136330.1 | - | 5 | 226 | |||
AK164299.1 | - | 5 | 232 | |||
BC016565.1 | - | 5 | 222 | |||
BC050116.1 | - | 5 | 221 | 6432881 | ||
BC098490.1 | - | 223 | 30022355 | |||
BY708305.1 | - | 5 | 244 | |||
CJ071503.1 | - | 5 | 290 |