Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028679.3 | NP_082955.2 | 5, 6 | 713 | ||
GenBank mRNA | AF461763.1 | - | 5, 6 | 693 | ||
AJ440757.2 | - | 5, 6 | 635 | |||
AK014783.1 | - | 5, 6 | 764 | |||
AK029057.1 | - | 5, 6 | 714 | |||
AK049210.1 | - | 5, 6 | 744 | |||
BC120829.1 | - | 5, 6 | 674 | 40129752 | ||
BC120831.1 | - | 5, 6 | 674 | 40129754 | ||
BC144835.1 | - | 5, 6 | 674 |