Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028711.3 | NP_082987.1 | 9 | 1559 | ||
GenBank mRNA | AB106930.1 | - | 9 | 1704 | ||
AK014394.1 | - | 9 | 1560 | |||
BC031371.1 | - | 1 | 350 | |||
BC138993.1 | - | 9 | 1566 | 8862015 | ||
BC138994.1 | - | 9 | 1566 | 8862016 |