Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159948.2 | NP_001153420.2 | 28, 29 | 4696 | ||
NM_028717.6 | NP_082993.4 | 28, 29 | 4527 | |||
GenBank mRNA | AB053307.1 | - | 28, 29 | 4515 | ||
AK014320.1 | - | 16, 17 | 1856 | |||
AK029858.1 | - | 28, 29 | 4700 | |||
AK171800.1 | - | 13, 14 | 1554 | |||
BC046828.1 | - | 28, 29 | 4488 | 6409924 |