Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000340.1 | NP_000331.1 | 4, 5 | 801 | ||
NM_001278658.1 | NP_001265587.1 | 3, 4 | 538 | |||
XM_011513087.2 | XP_011511389.1 | 5, 6 | 548 | |||
GenBank mRNA | AK290846.1 | - | 4, 5 | 544 | ||
AK292741.1 | - | 4, 5 | 544 | |||
AK298418.1 | - | 3, 4 | 302 | |||
AK313622.1 | - | 4, 5 | 542 | |||
J03810.1 | - | 4, 5 | 530 |