Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028721.3 | NP_082997.3 | 27 | 4053 | ||
GenBank mRNA | AK014393.1 | - | 12 | 1613 | ||
AK135256.1 | - | 2 | 146 | |||
AK173327.1 | - | 27 | 3632 | |||
AY259499.1 | - | 27 | 3990 | |||
BC115724.1 | - | 27 | 3580 | 40086844 |