Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028722.2 | NP_082998.2 | 7 | 1653 | ||
GenBank mRNA | AK014463.1 | - | 7 | 1906 | ||
AK015496.1 | - | 6 | 869 | |||
AK053719.1 | - | 6 | 1575 | |||
BC058629.1 | - | 7 | 1653 | 6402215 |