Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632428N05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159572.1 | NP_001153044.1 | 7 | 2652 | ||
NM_028732.4 | NP_083008.1 | 7 | 2655 | |||
GenBank mRNA | AK014600.1 | - | 7 | 2583 | ||
AK030479.1 | - | 7 | 2595 | |||
AK032383.1 | - | 7 | 2597 | |||
AK033393.1 | - | 7 | 2601 | |||
AK079734.1 | - | 7 | 2603 | |||
AK131118.1 | - | 7 | 2568 | |||
AK142494.1 | - | 1 | 501 | |||
AK163118.1 | - | 7 | 2600 | |||
AK170419.1 | - | 7 | 2600 | |||
AK171045.1 | - | 3 | 698 |