Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277292.1 | NP_001264221.1 | 22 | 3220 | ||
NM_001277293.1 | NP_001264222.1 | 23 | 3376 | |||
NM_001277294.1 | NP_001264223.1 | 22 | 3077 | |||
NM_001277295.1 | NP_001264224.1 | 12 | 1817 | |||
NM_028736.2 | NP_083012.1 | 24 | 3278 | |||
NM_130891.2 | NP_570961.1 | 21 | 2996 | |||
NM_133442.2 | NP_597699.1 | 21 | 3139 | |||
GenBank mRNA | AB051560.1 | - | 21 | 2873 | ||
AB051561.1 | - | 24 | 3155 | |||
AB051562.1 | - | 21 | 2859 | |||
AK016420.1 | - | 12 | 1815 | |||
AK029845.1 | - | 21 | 3538 | |||
AK029905.1 | - | 12 | 1818 | |||
AK220553.1 | - | 22 | 2934 | |||
AY255674.1 | - | 12 | 1633 | |||
AY294283.1 | - | 12 | 1614 | |||
BC067398.1 | - | 22 | 3077 | 30096759 | ||
BC072632.1 | - | 23 | 3376 | 6843802 |