Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000343.3 | NP_000334.1 | 3, 4 | 556 | ||
NM_001256314.1 | NP_001243243.1 | 2, 3 | 274 | |||
XM_011530331.1 | XP_011528633.1 | 3, 4 | 538 | |||
GenBank mRNA | AK297665.1 | - | 2, 3 | 248 | ||
AK312948.1 | - | 3, 4 | 460 | |||
AK316184.1 | - | 2, 3 | 253 | |||
BC139839.1 | - | 3, 4 | 325 | |||
CR456579.1 | - | 3, 4 | 356 | |||
DA429018.1 | - | 538 | ||||
M24847.1 | - | 3, 4 | 316 |