Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028773.3 | NP_083049.1 | 2 | 229 | ||
GenBank mRNA | AJ306422.1 | - | 2 | 95 | ||
AK004734.2 | - | 2 | 230 | |||
AK156202.1 | - | 2 | 294 | |||
AK171017.1 | - | 2 | 227 | |||
BC064734.1 | - | 2 | 214 |