Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028790.3 | NP_083066.1 | 2, 3 | 207 | ||
GenBank mRNA | AB078618.1 | - | 2, 3 | 204 | ||
AK004905.1 | - | 2, 3 | 208 | |||
AK033663.1 | - | 2, 3 | 207 | |||
AK034622.1 | - | 2, 3 | 211 | |||
BC025852.1 | - | 2, 3 | 160 | |||
BC132050.1 | - | 2, 3 | 238 | 40130327 | ||
BC137996.1 | - | 2, 3 | 238 | 8861015 |