Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145827.1 | NP_001139299.1 | 10 | 1421 | ||
NM_028800.3 | NP_083076.3 | 8 | 1067 | |||
GenBank mRNA | AB102946.1 | - | 8 | 898 | ||
AK009154.1 | - | 3 | 285 | |||
AK050808.1 | - | 4 | 370 | |||
AK154780.1 | - | 10 | 1412 | |||
AY336057.1 | - | 8 | 1052 | |||
BC046981.1 | - | 8 | 1026 | 6478265 |