Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 3, 4 | 566 | ||
XM_006716392.2 | XP_006716455.1 | 3, 4 | 749 | |||
GenBank mRNA | AF035277.1 | - | 3, 4 | 414 | ||
AK090513.1 | - | 2, 3 | 750 | |||
AK315122.1 | - | 3, 4 | 436 | |||
AL526126.3 | - | 3, 4 | 573 | |||
AV708514.1 | - | 1, 2 | 54 | |||
BC010550.1 | - | 3, 4 | 367 | 4153601 | ||
CR456803.1 | - | 3, 4 | 302 | |||
CR541850.1 | - | 3, 4 | 302 | |||
KU178337.1 | - | 3, 4 | 302 | |||
KU178338.1 | - | 3, 4 | 302 | |||
S79669.1 | - | 3, 4 | 454 | |||
U17280.1 | - | 3, 4 | 428 |