Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000351.5 | NP_000342.2 | 2 | 330 | ||
NM_001320750.1 | NP_001307679.1 | 3 | 554 | |||
NM_001320751.1 | NP_001307680.1 | 4 | 548 | |||
NM_001320752.1 | NP_001307681.1 | 3 | 394 | |||
NM_001320753.1 | NP_001307682.1 | 2 | 172 | |||
NM_001320754.1 | NP_001307683.1 | 2 | 320 | |||
GenBank mRNA | AK314034.1 | - | 2 | 270 | ||
AL549736.3 | - | 2 | 330 | |||
BC075030.2 | - | 2 | 106 | 30915417 | ||
DA327298.1 | - | 3 | 357 | |||
DA406759.1 | - | 3 | 395 | |||
J04964.1 | - | 2 | 288 | |||
M16505.1 | - | 2 | 305 |