Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028421.1 | NP_082697.1 | 2 | 746 | ||
NM_028864.2 | NP_083140.1 | 2 | 746 | |||
GenBank mRNA | AK004770.1 | - | 2 | 747 | ||
AK020263.2 | - | 2 | 425 | |||
AK080334.1 | - | 2 | 731 | |||
AK143568.1 | - | 2 | 416 | |||
AK169402.1 | - | 2 | 391 | |||
AK172379.1 | - | 2 | 747 | |||
BC029090.1 | - | 2 | 656 | 2651558 |