Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028865.3 | NP_083141.3 | 1, 2 | 268 | ||
GenBank mRNA | AK003450.1 | - | 1, 2 | 114 | ||
AK135294.1 | - | 229 | ||||
AK160133.1 | - | 1, 2 | 111 | |||
BC026936.1 | - | 1, 2 | 183 | 4021089 | ||
BY122329.1 | - | 1, 2 | 268 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK016897.1 | - | 229 | Chr. - - [-] on Build |