Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028868.3 | NP_083144.1 | 4 | 504 | ||
GenBank mRNA | AK010337.1 | - | 4 | 505 | ||
AK083655.1 | - | 4 | 509 | |||
AK146688.1 | - | 4 | 588 | |||
AY099096.1 | - | 4 | 476 | |||
BC030938.1 | - | 4 | 336 | 3980339 |