Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB082952.1 | - | 1 | 129 | ||
AK029341.1 | - | 1 | 925 | |||
AK087217.1 | - | 1 | 945 | |||
AK144082.1 | - | 1 | 924 | |||
AK161011.1 | - | 1 | 1035 | |||
AK172276.1 | - | 1 | 922 | |||
BC060140.1 | - | 1 | 241 | |||
BC118971.1 | - | 1 | 124 | |||
BC148309.2 | - | 1 | 129 |