Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000355.3 | NP_000346.2 | 2 | 461 | ||
NM_001184726.1 | NP_001171655.1 | 2 | 461 | |||
GenBank mRNA | AK225445.1 | - | 2 | 257 | ||
AK313711.1 | - | 2 | 351 | |||
BC001176.1 | - | 2 | 339 | 3353271 | ||
BC011239.1 | - | 2 | 340 | 4153362 | ||
CR456591.1 | - | 2 | 251 | |||
L02647.1 | - | 2 | 309 | |||
L02648.1 | - | 2 | 265 | |||
M60396.1 | - | 2 | 249 |