Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028906.2 | NP_083182.2 | 3 | 385 | ||
GenBank mRNA | AK016546.1 | - | 3 | 424 | ||
AK029788.1 | - | 2 | 2318 | |||
AK155605.1 | - | 3 | 429 | |||
BC043124.1 | - | 3 | 385 | 6410075 | ||
BC059222.1 | - | 3 | 385 | 6410075 |