Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930452B06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028934.3 | NP_083210.3 | 3 | 454 | ||
GenBank mRNA | AK016701.1 | - | 3 | 403 | ||
AK019627.1 | - | 3 | 411 | |||
AK047451.1 | - | 3 | 472 | |||
AK050783.1 | - | 3 | 438 | |||
BC064468.1 | - | 3 | 404 | 6333742 |