Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028946.3 | NP_083222.2 | 4 | 662 | ||
GenBank mRNA | AK007064.1 | - | 4 | 654 | ||
AK015318.1 | - | 4 | 629 | |||
AK016883.1 | - | 4 | 651 | |||
AK016917.1 | - | 4 | 642 | |||
AK029525.1 | - | 4 | 663 | |||
BC025002.1 | - | 3 | 584 | |||
EU846100.1 | - | 3 | 193 |