Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028756.4 | NP_083032.2 | 6 | 946 | ||
GenBank mRNA | AK003124.1 | - | 6 | 874 | ||
AK030566.1 | - | 6 | 871 | |||
AK033921.1 | - | 6 | 913 | |||
AK085567.1 | - | 6 | 885 | |||
AK150825.1 | - | 6 | 937 | |||
AK151484.1 | - | 6 | 937 | |||
AK170026.1 | - | 6 | 913 | |||
BC011115.1 | - | 6 | 875 | 4013987 |