Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166138.1 | NP_001159610.1 | 2, 3 | 376 | ||
NM_028768.3 | NP_083044.2 | 2, 3 | 376 | |||
GenBank mRNA | AK004793.2 | - | 2, 3 | 377 | ||
AK078280.1 | - | 2, 3 | 127 | |||
BC038866.1 | - | 2, 3 | 334 | 1230321 |