Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028771.2 | NP_083047.1 | 2, 3 | 597 | ||
GenBank mRNA | AK004762.1 | - | 2, 3 | 598 | ||
AK013304.1 | - | 2, 3 | 548 | |||
AK045509.1 | - | 2, 3 | 589 | |||
AK088221.1 | - | 2, 3 | 588 | |||
AK155466.1 | - | 2, 3 | 580 | |||
BC006752.1 | - | 1, 2 | 102 | |||
BC078634.1 | - | 2, 3 | 564 | 5388281 |