Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286002.1 | NP_001272931.1 | 2, 3 | 449 | ||
NM_028776.5 | NP_083052.1 | 2, 3 | 636 | |||
GenBank mRNA | AK004809.2 | - | 2, 3 | 376 | ||
AK030824.1 | - | 2, 3 | 612 | |||
AK162937.1 | - | 2, 3 | 450 | |||
BC043085.1 | - | 2, 3 | 337 | 6417440 | ||
BC066800.1 | - | 2, 3 | 321 |