Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195025.1 | NP_001181954.1 | 2 | 421 | ||
NM_028778.4 | NP_083054.1 | 2 | 421 | |||
GenBank mRNA | AK004737.2 | - | 2 | 409 | ||
AK033672.1 | - | 2 | 408 | |||
AK034082.1 | - | 2 | 413 | |||
AK164419.1 | - | 1 | 3218 | |||
BC033302.1 | - | 2 | 396 | 3663595 | ||
BC046833.1 | - | 2 | 396 | 6816569 |