Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004444.1 | - | 2 | 288 | ||
AK076500.1 | - | 2 | 275 | |||
AK151245.1 | - | 1 | 95 | |||
AK158058.1 | - | 2 | 350 | |||
AK206364.1 | - | 1 | 50 | |||
BC033921.1 | - | 2 | 260 | 5068809 | ||
BY237462.1 | - | 2 | 399 |