Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170966.1 | NP_001164437.1 | 5 | 620 | ||
NM_001170967.1 | NP_001164438.1 | 5 | 620 | |||
NM_001170970.1 | NP_001164441.1 | 5 | 620 | |||
NM_028866.3 | NP_083142.2 | 5 | 620 | |||
GenBank mRNA | AB086191.1 | - | 4 | 399 | ||
AK003256.1 | - | 5 | 588 | |||
AK009297.1 | - | 5 | 587 | |||
AK045125.1 | - | 5 | 580 | |||
AK045923.1 | - | 5 | 579 | |||
AK050653.1 | - | 5 | 580 | |||
AK078286.1 | - | 5 | 588 | |||
AK161982.1 | - | 5 | 580 | |||
BC011156.1 | - | 5 | 532 | |||
BC019463.1 | - | 5 | 620 | |||
BC030076.1 | - | 5 | 552 | |||
BC064745.1 | - | 5 | 501 |