Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028870.3 | NP_083146.1 | 5 | 1028 | ||
GenBank mRNA | AK029382.1 | - | 5 | 1029 | ||
AK146816.1 | - | 1101 | ||||
AK159480.1 | - | 5 | 1078 | |||
AK167531.1 | - | 1101 | ||||
AK213322.1 | - | 1 | 69 |