Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028873.4 | NP_083149.3 | 2 | 402 | ||
GenBank mRNA | AK017830.1 | - | 2 | 416 | ||
AK032681.1 | - | 2 | 383 | |||
AK045345.1 | - | 2 | 459 | |||
AK045445.1 | - | 2 | 496 | |||
AK053314.1 | - | 2 | 464 | |||
AK082579.1 | - | 2 | 403 | |||
AK083878.1 | - | 2 | 413 | |||
AK159388.1 | - | 2 | 415 | |||
AK168935.1 | - | 2 | 493 | |||
AK198987.1 | - | 1 | 147 | |||
BC011146.1 | - | 2 | 467 | 4016209 |