Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028887.3 | NP_083163.3 | 27 | 3663 | ||
GenBank mRNA | AK016419.1 | - | 2 | 140 | ||
AK129181.1 | - | 5 | 660 | |||
BC058618.1 | - | 3 | 315 | |||
BC062946.1 | - | 13 | 1508 | 30357469 | ||
BC076627.1 | - | 20 | 2626 |