Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB082952.1 | - | 1, 2 | 164 | ||
AK029341.1 | - | 1, 2 | 960 | |||
AK087217.1 | - | 1, 2 | 980 | |||
AK144082.1 | - | 1, 2 | 959 | |||
AK161011.1 | - | 1, 2 | 1070 | |||
AK172276.1 | - | 1, 2 | 957 | |||
BC060140.1 | - | 1, 2 | 276 | |||
BC118971.1 | - | 1, 2 | 159 | |||
BC148309.2 | - | 1, 2 | 164 |