Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930452B06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028934.3 | NP_083210.3 | 3, 4 | 489 | ||
GenBank mRNA | AK016701.1 | - | 3, 4 | 438 | ||
AK019627.1 | - | 3, 4 | 446 | |||
AK050783.1 | - | 3, 4 | 473 | |||
AK133893.1 | - | 3, 4 | 491 | |||
BC064468.1 | - | 3, 4 | 439 | 6333742 |