Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028946.3 | NP_083222.2 | 4 | 677 | ||
GenBank mRNA | AK007064.1 | - | 4 | 669 | ||
AK015318.1 | - | 4 | 644 | |||
AK016883.1 | - | 4 | 666 | |||
AK016917.1 | - | 4 | 657 | |||
AK029525.1 | - | 4 | 678 | |||
BC025002.1 | - | 3 | 599 | |||
EU846100.1 | - | 3 | 208 |