Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028763.3 | NP_083039.2 | 5 | 3193 | ||
GenBank mRNA | AK004679.2 | - | 5 | 3098 | ||
AK035250.1 | - | 5 | 3100 | |||
AK090253.1 | - | 5 | 3048 | |||
AK146048.1 | - | 5 | 3044 | |||
AK146103.1 | - | 5 | 3044 | |||
AK147702.1 | - | 1 | 7796 | |||
AK162544.1 | - | 1 | 193 | |||
AK202886.1 | - | 1 | 333 | |||
AK207037.1 | - | 1 | 361 | |||
AK218930.1 | - | 1 | 355 | |||
BC011199.1 | - | 5 | 3082 | |||
BC048240.1 | - | 5 | 3090 | 6493113 |