Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028771.2 | NP_083047.1 | 5 | 2516 | ||
GenBank mRNA | AK004762.1 | - | 5 | 2517 | ||
AK088221.1 | - | 5 | 2507 | |||
AK155466.1 | - | 5 | 2500 | |||
AK177314.1 | - | 1 | 122 | |||
AK208547.1 | - | 1 | 157 | |||
BC006752.1 | - | 4 | 2021 | |||
BC078634.1 | - | 5 | 2483 | 5388281 |