Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028773.3 | NP_083049.1 | 8 | 2359 | ||
GenBank mRNA | AK004734.2 | - | 8 | 2360 | ||
AK156202.1 | - | 8 | 2424 | |||
AK171017.1 | - | 8 | 2357 | |||
BC028773.1 | - | 5 | 1921 | |||
BC064734.1 | - | 8 | 2344 |