Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000373.3 | NP_000364.1 | 2 | 307 | ||
NR_033437.1 | 3 | 406 | ||||
GenBank mRNA | AB062285.1 | - | 2 | 229 | ||
AK291306.1 | - | 2 | 223 | |||
BC000364.2 | - | 2 | 224 | 2964556 | ||
BC007511.2 | - | 2 | 224 | 2964556 | ||
CR456787.1 | - | 2 | 201 | |||
D86227.1 | - | 2 | 220 | |||
D86228.1 | - | 2 | 220 | |||
D86230.1 | - | 2 | 220 | |||
EU921886.1 | - | 2 | 221 | |||
EU921890.1 | - | 2 | 221 | |||
EU921891.1 | - | 3 | 320 | |||
EU921892.1 | - | 2 | 221 | |||
EU921893.1 | - | 2 | 221 | |||
EU921894.1 | - | 2 | 221 | |||
J03626.1 | - | 2 | 305 | |||
KU178375.1 | - | 2 | 201 | |||
KU178376.1 | - | 3 | 300 | |||
M36661.1 | - | 2 | 167 |